Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179638.2 | NP_849969.2 | 13 - 14 | 1439 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |