Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179659.2 | NP_849990.1 | 18 - 19 | 1913 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |