Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179659.2 | NP_849990.1 | 7 - 8 | 954 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |