Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124889.2 | NP_001118361.1 | 13 - 14 | 2955 | 152 | |
NM_001335795.1 | NP_001325089.1 | 12 - 13 | 3481 | 152 | ||
NM_179692.3 | NP_850023.1 | 12 - 13 | 3027 | 152 | ||
GenBank mRNA | AF506697.1 | - | 12 - 13 | 2651 | 152 | |
AY510025.1 | - | 12 - 13 | 2651 | 152 | ||
BT002459.1 | - | 12 - 13 | 2834 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |