Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OVA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336021.1 | NP_001323723.1 | 1 - 2 | 263 | 84 | |
NM_179739.4 | NP_850070.1 | 1 - 2 | 419 | 84 | ||
NM_201804.3 | NP_973533.1 | 1 - 2 | 263 | 84 | ||
NM_201805.1 | NP_973534.1 | 1 - 2 | 206 | 84 | ||
GenBank mRNA | AY081299.1 | - | 1 - 2 | 206 | 84 | |
BT001221.1 | - | 1 - 2 | 187 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |