Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179749.3 | NP_850080.1 | 1 - 2 | 345 | 69 | |
NM_179750.3 | NP_850081.1 | 2 - 3 | 188 | 69 | ||
GenBank mRNA | AK119091.1 | - | 1 - 2 | 114 | 69 | |
BT003734.1 | - | 1 - 2 | 74 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Arabidopsis thaliana Probe spans exons |