Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036358.2 | NP_001031435.1 | 6 - 7 | 1409 | 75 | |
NM_001336153.1 | NP_001323735.1 | 6 - 7 | 1142 | 75 | ||
NM_001336154.1 | NP_001323736.1 | 6 - 7 | 883 | 75 | ||
NM_001336155.1 | NP_001323737.1 | 6 - 7 | 1268 | 75 | ||
NM_001336156.1 | NP_001323738.1 | 6 - 7 | 1191 | 75 | ||
NM_179784.2 | NP_850115.1 | 5 - 6 | 1378 | 75 | ||
GenBank mRNA | AY065457.1 | - | 5 - 6 | 930 | 75 | |
BT004417.1 | - | 5 - 6 | 760 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |