Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084502.2 | NP_001077971.1 | 4 - 5 | 404 | 69 | |
NM_001336158.1 | NP_001325347.1 | 3 - 4 | 290 | 69 | ||
NM_001336159.1 | NP_001325348.1 | 3 - 4 | 290 | 69 | ||
NM_001336160.1 | NP_001325349.1 | 3 - 4 | 290 | 69 | ||
NM_179785.3 | NP_850116.1 | 4 - 5 | 404 | 69 | ||
NM_179786.2 | NP_850117.1 | 4 - 5 | 404 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |