Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftsh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179798.2 | NP_850129.1 | 2 - 3 | 600 | 147 | |
GenBank mRNA | BT002743.1 | - | 2 - 3 | 270 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |