Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT4;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128519.6 | NP_180526.1 | 1 - 2 | 432 | 94 | |
NM_179805.2 | NP_850136.1 | 1 - 2 | 447 | 94 | ||
GenBank mRNA | BT001983.1 | - | 1 - 2 | 406 | 94 | |
BT008489.1 | - | 1 - 2 | 358 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |