Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHT4;1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336220.1 | NP_001325344.1 | 7 - 8 | 1558 | 62 | |
NM_128519.6 | NP_180526.1 | 8 - 9 | 1511 | 62 | ||
NM_179805.2 | NP_850136.1 | 8 - 9 | 1545 | 62 | ||
NM_201832.1 | NP_973561.1 | 8 - 9 | 1324 | 62 | ||
GenBank mRNA | BT001983.1 | - | 8 - 9 | 1504 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |