Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179819.2 | NP_850150.1 | 6 - 7 | 990 | 78 | |
GenBank mRNA | AY081313.1 | - | 6 - 7 | 983 | 78 | |
BT002563.1 | - | 6 - 7 | 678 | 78 | ||
KJ138886.1 | - | 6 - 7 | 678 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |