Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179845.4 | NP_850176.1 | 3 - 4 | 2030 | 67 | |
GenBank mRNA | AY065010.1 | - | 3 - 4 | 1917 | 67 | |
AY129492.1 | - | 3 - 4 | 1865 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |