Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036384.2 | NP_001031461.1 | 3 - 4 | 787 | 129 | |
NM_001202723.1 | NP_001189652.1 | 2 - 3 | 798 | 129 | ||
NM_001336357.1 | NP_001324733.1 | 2 - 3 | 880 | 129 | ||
NM_001336358.1 | NP_001324731.1 | 3 - 4 | 813 | 129 | ||
NM_001336359.1 | NP_001324732.1 | 3 - 4 | 778 | 129 | ||
NM_179846.4 | NP_850177.2 | 2 - 3 | 880 | 129 | ||
GenBank mRNA | AY065265.1 | - | 2 - 3 | 762 | 129 | |
AY096557.1 | - | 2 - 3 | 661 | 129 | ||
Z48047.1 | - | 784 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |