Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036384.2 | NP_001031461.1 | 4 - 5 | 955 | 127 | |
NM_001202723.1 | NP_001189652.1 | 3 - 4 | 966 | 127 | ||
NM_001336357.1 | NP_001324733.1 | 3 - 4 | 1048 | 127 | ||
NM_001336358.1 | NP_001324731.1 | 4 - 5 | 981 | 127 | ||
NM_001336359.1 | NP_001324732.1 | 4 - 5 | 946 | 127 | ||
NM_179846.4 | NP_850177.2 | 3 - 4 | 1048 | 127 | ||
GenBank mRNA | AY065265.1 | - | 3 - 4 | 930 | 127 | |
AY096557.1 | - | 3 - 4 | 829 | 127 | ||
Z48047.1 | - | 952 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |