Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112402.3 | NP_188154.1 | 2 - 3 | 448 | 138 | |
GenBank mRNA | AF361806.1 | - | 2 - 3 | 350 | 138 | |
AY059155.1 | - | 2 - 3 | 231 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |