Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202971.2 | NP_001189900.1 | 7 - 8 | 755 | 84 | |
NM_001338166.1 | NP_001327652.1 | 8 - 9 | 1364 | 84 | ||
NM_112407.5 | NP_188158.4 | 9 - 10 | 1409 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |