Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035626.2 | NP_001030703.1 | 4 - 5 | 784 | 77 | |
NM_112432.2 | NP_566520.1 | 4 - 5 | 959 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |