Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035626.2 | NP_001030703.1 | 9 - 10 | 1207 | 76 | |
NM_112432.2 | NP_566520.1 | 9 - 10 | 1382 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |