Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035627.3 | NP_001030704.1 | 3 - 4 | 381 | 84 | |
NM_112436.3 | NP_566522.1 | 2 - 3 | 458 | 84 | ||
GenBank mRNA | AK227628.1 | - | 2 - 3 | 327 | 84 | |
AK317312.1 | - | 3 - 4 | 314 | 84 | ||
BT006396.1 | - | 2 - 3 | 217 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |