Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035631.2 | NP_001030708.1 | 10 - 11 | 694 | 76 | |
NM_001338191.1 | NP_001326809.1 | 10 - 11 | 694 | 76 | ||
NM_112465.4 | NP_188216.2 | 12 - 13 | 808 | 76 | ||
GenBank mRNA | AY120750.1 | - | 12 - 13 | 807 | 76 | |
BT001207.1 | - | 12 - 13 | 766 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |