Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035631.2 | NP_001030708.1 | 21 - 22 | 1312 | 128 | |
NM_001338192.1 | NP_001326807.1 | 15 - 16 | 964 | 128 | ||
NM_001338193.1 | NP_001326808.1 | 13 - 14 | 851 | 128 | ||
NM_112465.4 | NP_188216.2 | 23 - 24 | 1426 | 128 | ||
GenBank mRNA | AY120750.1 | - | 23 - 24 | 1425 | 128 | |
BT001207.1 | - | 23 - 24 | 1384 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |