Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDIL1-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112481.4 | NP_188232.2 | 10 - 11 | 1692 | 79 | |
GenBank mRNA | BT015349.1 | - | 10 - 11 | 1514 | 79 | |
BT015798.1 | - | 10 - 11 | 1459 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |