Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202983.2 | NP_001189912.1 | 4 - 5 | 473 | 67 | |
NM_112507.1 | NP_566544.1 | 3 - 4 | 311 | 67 | ||
GenBank mRNA | AB041766.1 | - | 3 - 4 | 311 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |