Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338255.1 | NP_001326711.1 | 15 - 16 | 3121 | 97 | |
NM_001338256.1 | NP_001326710.1 | 15 - 16 | 3121 | 97 | ||
NM_001338257.1 | NP_001326709.1 | 15 - 16 | 3075 | 97 | ||
NM_001338258.1 | NP_001326708.1 | 15 - 16 | 3060 | 97 | ||
NM_001338259.1 | NP_001326707.1 | 15 - 16 | 3060 | 97 | ||
NM_112553.4 | NP_188302.2 | 15 - 16 | 3121 | 97 | ||
GenBank mRNA | AF411833.1 | - | 15 - 16 | 2928 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |