Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338286.1 | NP_001326050.1 | 23 - 24 | 5183 | 159 | |
NM_001338287.1 | NP_001326049.1 | 23 - 24 | 5142 | 159 | ||
NM_112614.4 | NP_188362.2 | 23 - 24 | 5159 | 159 | ||
GenBank mRNA | DQ399529.1 | - | 23 - 24 | 4931 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |