Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112621.6 | NP_188368.2 | 5 - 6 | 1466 | 84 | |
GenBank mRNA | AY065430.1 | - | 5 - 6 | 1331 | 84 | |
AY096380.1 | - | 4 - 5 | 1095 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |