Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPSN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035639.2 | NP_001030716.1 | 9 - 10 | 928 | 84 | |
NM_112623.5 | NP_566578.1 | 9 - 10 | 994 | 84 | ||
GenBank mRNA | AY099622.1 | - | 9 - 10 | 862 | 84 | |
BT001249.1 | - | 9 - 10 | 712 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |