Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112626.3 | NP_566579.1 | 2 - 3 | 708 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |