Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202989.1 | NP_001189918.1 | 4 - 5 | 391 | 95 | |
NM_001338294.1 | NP_001326663.1 | 4 - 5 | 443 | 95 | ||
NM_112639.4 | NP_566581.1 | 4 - 5 | 443 | 95 | ||
GenBank mRNA | AK117162.1 | - | 4 - 5 | 385 | 95 | |
BT024620.1 | - | 4 - 5 | 310 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |