Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112651.5 | NP_566585.1 | 10 - 11 | 2273 | 61 | |
GenBank mRNA | AF148541.1 | - | 10 - 11 | 2152 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |