Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338307.1 | NP_001326341.1 | 5 - 6 | 1333 | 66 | |
NM_112651.5 | NP_566585.1 | 5 - 6 | 1431 | 66 | ||
GenBank mRNA | AF148541.1 | - | 5 - 6 | 1310 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |