Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOC64-III | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338317.1 | NP_001325422.1 | 6 - 7 | 1020 | 74 | |
NM_112678.4 | NP_188424.2 | 6 - 7 | 1065 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |