Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HON4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338318.1 | NP_001325770.1 | 3 - 4 | 2064 | 86 | |
NM_112685.4 | NP_188431.3 | 3 - 4 | 1609 | 86 | ||
GenBank mRNA | BT002434.1 | - | 3 - 4 | 1332 | 86 | |
BT006606.1 | - | 3 - 4 | 1162 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |