Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112749.5 | NP_188493.1 | 5 - 6 | 1031 | 106 | |
GenBank mRNA | AY084956.1 | - | 5 - 6 | 903 | 106 | |
BT029175.1 | - | 5 - 6 | 837 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |