Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112759.5 | NP_188503.2 | 14 - 15 | 3191 | 66 | |
GenBank mRNA | AB112744.1 | - | 14 - 15 | 3149 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |