Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112791.6 | NP_188535.4 | 20 - 21 | 2650 | 75 | |
GenBank mRNA | DQ399530.1 | - | 19 - 20 | 2450 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |