Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338383.1 | NP_001325693.1 | 2 - 3 | 912 | 80 | |
NM_112802.4 | NP_188546.1 | 3 - 4 | 820 | 80 | ||
GenBank mRNA | AF106705.2 | - | 3 - 4 | 546 | 80 | |
AJ301557.1 | - | 3 - 4 | 524 | 80 | ||
BT030623.1 | - | 3 - 4 | 524 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |