Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035648.1 | NP_001030725.1 | 10 - 11 | 2609 | 105 | |
NM_112805.5 | NP_188549.2 | 11 - 12 | 2541 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |