Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203004.1 | NP_001189933.1 | 20 - 21 | 2531 | 72 | |
NM_112808.4 | NP_188552.3 | 20 - 21 | 2512 | 72 | ||
GenBank mRNA | AB250666.1 | - | 20 - 21 | 2487 | 72 | |
DQ912973.1 | - | 20 - 21 | 2425 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |