Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203005.2 | NP_001189934.1 | 2 - 3 | 1381 | 66 | |
NM_001338390.1 | NP_001326148.1 | 2 - 3 | 1312 | 66 | ||
NM_001338391.1 | NP_001326147.1 | 1 - 2 | 1835 | 66 | ||
NM_112816.3 | NP_566629.1 | 2 - 3 | 1317 | 66 | ||
GenBank mRNA | AB017161.1 | - | 1 - 2 | 1153 | 66 | |
AF093547.1 | - | 1 - 2 | 1187 | 66 | ||
AY050897.1 | - | 2 - 3 | 1283 | 66 | ||
AY091298.1 | - | 1 - 2 | 1116 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |