Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112861.4 | NP_188605.1 | 2 - 3 | 359 | 63 | |
GenBank mRNA | AF446892.1 | - | 2 - 3 | 206 | 63 | |
AJ271732.1 | - | 2 - 3 | 255 | 63 | ||
AK226525.1 | - | 2 - 3 | 275 | 63 | ||
AY052676.1 | - | 2 - 3 | 262 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |