Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOM40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338439.1 | NP_001326561.1 | 5 - 6 | 873 | 96 | |
NM_112890.5 | NP_188634.1 | 5 - 6 | 681 | 96 | ||
GenBank mRNA | AY050925.1 | - | 5 - 6 | 523 | 96 | |
AY117216.1 | - | 5 - 6 | 458 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |