Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DELTA-VPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112912.3 | NP_188656.1 | 3 - 4 | 649 | 136 | |
GenBank mRNA | AB105106.1 | - | 3 - 4 | 550 | 136 | |
AF521661.1 | - | 3 - 4 | 553 | 136 | ||
AY120765.1 | - | 3 - 4 | 576 | 136 | ||
BT000949.1 | - | 3 - 4 | 514 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |