Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035654.2 | NP_001030731.1 | 5 - 6 | 474 | 94 | |
NM_001338474.1 | NP_001327849.1 | 6 - 7 | 552 | 94 | ||
NM_112920.4 | NP_566657.1 | 6 - 7 | 619 | 94 | ||
GenBank mRNA | AK317308.1 | - | 5 - 6 | 540 | 94 | |
AY034911.1 | - | 6 - 7 | 476 | 94 | ||
BT004414.1 | - | 5 - 6 | 346 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |