Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338479.1 | NP_001327803.1 | 2 - 3 | 935 | 81 | |
NM_112932.3 | NP_188676.1 | 1 - 2 | 1297 | 81 | ||
GenBank mRNA | AY120727.1 | - | 1 - 2 | 915 | 81 | |
BT008898.1 | - | 1 - 2 | 728 | 81 | ||
U31751.1 | - | 1 - 2 | 802 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |