Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112942.5 | NP_188686.2 | 1 - 2 | 173 | 93 | |
GenBank mRNA | AY062488.1 | - | 1 - 2 | 162 | 93 | |
AY093272.1 | - | 1 - 2 | 147 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |