Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLUR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342335.1 | NP_001320141.1 | 4 - 5 | 2409 | 99 | |
NM_001342336.1 | NP_001329586.1 | 5 - 6 | 2203 | 99 | ||
NM_001342337.1 | NP_001329588.1 | 5 - 6 | 2113 | 99 | ||
NM_001342338.1 | NP_001329587.1 | 5 - 6 | 2129 | 99 | ||
NM_119695.4 | NP_567981.1 | 4 - 5 | 2035 | 99 | ||
NM_202957.3 | NP_974686.1 | 5 - 6 | 2203 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |