Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119836.3 | NP_195390.2 | 1 - 2 | 130 | 111 | |
GenBank mRNA | BT012190.1 | - | 1 - 2 | 70 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |