Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEJ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119855.4 | NP_195409.1 | 7 - 8 | 900 | 66 | |
GenBank mRNA | AY085164.1 | - | 7 - 8 | 729 | 66 | |
BT003835.1 | - | 7 - 8 | 715 | 66 | ||
BT005190.1 | - | 7 - 8 | 639 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |